Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPSD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562007
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562007 Supplier Invitrogen™ Supplier No. PA562007
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KDLAALRVQL REQHLYYQDQ LLPVSRIIV Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 59%.

Tryptases comprise a family of trypsin-like serine proteases, the peptidase family S1. Tryptases are enzymatically active only as heparin-stabilized tetramers, and they are resistant to all known endogenous proteinase inhibitors. Several tryptase genes are clustered on chromosome 16p13.3. These genes are characterized by several distinct features. They have a highly conserved 3' UTR and contain tandem repeat sequences at the 5' flank and 3' UTR which are thought to play a role in regulation of the mRNA stability. Although this gene may be an exception, most of the tryptase genes have an intron immediately upstream of the initiator Met codon, which separates the site of transcription initiation from protein coding sequence. This feature is characteristic of tryptases but is unusual in other genes. Tryptases have been implicated as mediators in the pathogenesis of asthma and other allergic and inflammatory disorders. TPSD1 was once considered to be a pseudogene, although it is now believed to be a functional gene that encodes a protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPSD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TPSD1
Gene Accession No. Q9BZJ3
Gene Alias Delta-tryptase; hmMCP-3-like tryptase III; Mast cell mMCP-7-like; mast cell tryptase; MCP7L1; MCP7-LIKE; MMCP-7L; mMCP-7-like delta II tryptase; mMCP-7-like-1; mMCP-7-like-2; TPSD1; Tryptase delta; tryptase delta 1; tryptase-3
Gene Symbols TPSD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TPSD1. Recombinant protein control fragment (Product #RP-104454).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23430
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.