Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPSG1 Monoclonal Antibody (OTI3F2), TrueMAB™, OriGene
SDP

Catalog No. 502540547
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0547 30 μL
50-167-4803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0547 Supplier OriGene Supplier No. TA504030S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TPSG1 Monoclonal antibody specifically detects TPSG1 in Human samples. It is validated for Western Blot

Specifications

Antigen TPSG1
Applications Western Blot
Classification Monoclonal
Clone OTI3F2
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TPSG1
Gene Accession No. Q9NRR2
Gene Alias PRSS31; serine protease 31; TMT; TPSG1; Transmembrane tryptase; trpA; Tryptase gamma; Tryptase- gamma; tryptase gamma 1; Tryptase gamma heavy chain; tryptase gamma I; tryptase gamma II; Tryptase gamma light chain
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 20-283 of human TPSG1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25823
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.