Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPST1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572357
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572357 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572357 Supplier Invitrogen™ Supplier No. PA572357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TPST1 Polyclonal Antibody for Western Blot

The tyrosylprotein sulfotransferases TPST-1 and TPST-2 catalyze the sulfation of tyrosine residues within secreted and membrane-bound proteins, such as cell adhesion molecules, G-protein-coupled receptors, coagulation factors, serpins, extracellular matrix proteins, and hormones. Although both TPST-1 and TPST-2 utilize 3'-phosphoadenosine 5'-phosphosulfate as their sulfate donor, they differ in their substrate specificity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPST1
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TPST1
Gene Accession No. O60507, O70281
Gene Alias Protein-tyrosine sulfotransferase 1; R75054; TANGO13A; Tpst1; TPST-1; transport and golgi organization 13 homolog A; tyrosylprotein sulfotransferase 1; tyrosylprotein sulfotransferase-1
Gene Symbols TPST1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 336-366 amino acids from the C-terminal region of human TPST1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22021, 8460
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.