Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPST2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554319 Supplier Invitrogen™ Supplier No. PA554319
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IAKHGEPARV LCNKDPFTLK SSVYLSRLFP NSKFLLMVRD GRASVHSMIT RKVTIAGFDL SSYRDCLTKW NKAIEVMYAQ CMEVGKEKCL PVYYEQLVLH PRRSLKLILD FLGIAWSDAV LHHEDLIGKP GGVSLSKIER STDQVIKPVN LEALSKWTGH IPGDVVRDMA QIAPMLAQLG Y Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

The ADP-ribosylation factor (Arf) family are highly conserved members of the Ras superfamily of regulatory GTP-binding proteins. Arf proteins participate in routing of intracellular proteins to and within the Golgi complex. Cellular functions include maintenance of organelle integrity, coat protein assembly, as an activator of phospholipase D. The Arf family is divided functionally into the Arf and the Arf-like (Arl) proteins. The ARF proteins are categorized as class I (ARF1, ARF2, and ARF3), class II (ARF4 and ARF5) and class III (ARF6) and members of each class share a common gene organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPST2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tpst2
Gene Accession No. O60704
Gene Alias AI448750; D5ucla3; grm; grt; hypothetical protein LOC540183; protein-tyrosine sulfotransferase 2; Tango13b; Tpst2; TPST-2; transport and golgi organization 13 homolog B; tyrosylprotein phosphotransferase 2; tyrosylprotein sulfotransferase 2; tyrosylprotein sulfotransferase-2
Gene Symbols Tpst2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TPST2. Recombinant protein control fragment (Product #RP-91339).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8459
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.