Learn More
Invitrogen™ TPT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528273
Description
Recommended positive controls: mouse brain. Predicted reactivity: Mouse (95%), Rat (95%), Pig (100%), Rabbit (98%), Chicken (90%), Sheep (99%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (99%), Guinea pig (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TPT1 (translationally controlled tumor protein 1) is a 23 kDa member of the TCTP family of calcium binding proteins. TPT1 is localized in the cytoplasm and widely expressed and serves as a transcriptional activator, calcium transporter, histamine inducer and antiapoptotic caspase 3 inhibitor. TPT1 is a cytokine-like molecule that causes the release of histamine, IL-4 and IL-13 from basophils as well as the secretion of IL-8 and a calcium response in eosinophils. TPT1 plays a pivotal role in allergic diseases and due to its wide distribution in brain, is thought to be involved in neurodegenerative disorders, such as Alzheimer's disease and Down syndrome.
Specifications
| TPT1 | |
| Polyclonal | |
| Unconjugated | |
| TPT1 | |
| 21 kDa polypeptide; 21 kDa polypeptide under translational control; fortilin; Histamine-releasing factor; HRF; Lens epithelial protein; p02; p21; p23; TCTP; Tpt1; translationally regulated transcript (21 kDa); translationally-controlled tumor protein; Trt; tumor protein, translationally-controlled 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22070, 7178 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.54 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P13693, P63028 | |
| TPT1 | |
| Recombinant protein encompassing a sequence within the center region of human TCTP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.