Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPT1/TCTP Antibody (10A9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | TPT1/TCTP |
---|---|
Clone | 10A9 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry |
Classification | Monoclonal |
Description
TPT1/TCTP Monoclonal antibody specifically detects TPT1/TCTP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, ImmunocytochemistrySpecifications
TPT1/TCTP | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
FLJ27337, Fortilin, Histamine-releasing factor, p02, p23, TCTPHRFfortilin, translationally-controlled tumor protein, tumor protein, translationally-controlled 1 | |
A synthesized peptide derived from Human TPT1/TCTP [UniProt P13693] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
10A9 | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
7178 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title