Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPT1/TCTP Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | TPT1/TCTP |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TPT1/TCTP Polyclonal specifically detects TPT1/TCTP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TPT1/TCTP | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
FLJ27337, Fortilin, Histamine-releasing factor, p02, p23, TCTPHRFfortilin, translationally-controlled tumor protein, tumor protein, translationally-controlled 1 | |
TPT1 | |
IgG | |
Affinity Purified |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Stem Cell Markers | |
PBS with 0.02% Sodium Azide | |
7178 | |
Antibody was raised against a 19 amino acid peptide near the center of human TPT1. The immunogen is located within amino acids 80 - 130 of TPT1. Amino Acid Squence: DYMKSIKGKLEEQRPERVK | |
Primary | |
19 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title