Learn More
Invitrogen™ TPTE2 Monoclonal Antibody (978CT6.1.4)

Description
TPTE2 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | TPTE2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 978CT6.1.4 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.09% sodium azide; pH 7.4 |
| Gene | TPTE2 |
| Gene Accession No. | Q6XPS3 |
| Gene Alias | Lipid phosphatase TPIP; phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase TPTE2; phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase TPTE2; PTEN-like inositol lipid phosphatase; TPIP; TPIP lipid phosphatase; TPTE and PTEN homologous inositol lipid phosphatase; TPTE2; transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.