Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPTE2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566988
Description
Immunogen sequence: ERAYDPKHFHN RVSRIMIDDH NV Highest antigen sequence identity to the following orthologs - mouse 91%, rat 83%.
TPIP is a member of a large class of membrane-associated phosphatases with substrate specificity for the 3-position phosphate of inositol phospholipids.[supplied by OMIM, Jul 2002].
Specifications
TPTE2 | |
Polyclonal | |
Unconjugated | |
TPTE2 | |
Lipid phosphatase TPIP; phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase TPTE2; phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase TPTE2; PTEN-like inositol lipid phosphatase; TPIP; TPIP lipid phosphatase; TPTE and PTEN homologous inositol lipid phosphatase; TPTE2; transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
93492 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6XPS3 | |
TPTE2 | |
Recombinant Human TPTE2. Recombinant protein control fragment (Product #RP-105652). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction