Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPX2 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500183C
Description
TPX2 Polyclonal antibody specifically detects TPX2 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
TPX2 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
C20orf1HCTP4, C20orf2, chromosome 20 open reading frame 1, Differentially expressed in cancerous and non-cancerous lung cells 2, differentially expressed in lung cells, DIL2, DIL-2FLS353, HCA519, Hepatocellular carcinoma-associated antigen 519, P100, p100GD:C20orf1, preferentially expressed in colorectal cancer, Protein fls353, REPP86, restricted expression proliferation associated protein 100, Restricted expression proliferation-associated protein 100, targeting protein for Xklp2, TPX2, microtubule-associated protein homolog, TPX2, microtubule-associated, homolog (Xenopus laevis) | |
The immunogen recognized by this antibody maps to a region between residues 150 and 200 of human TPX2 using the numbering given in Swiss-Prot entry Q9ULW0 (GeneID 22974). | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Mitotic Regulators | |
22974 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction