Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPX2 Mouse anti-Human, Clone: 18D5, Abnova™

Catalog No. 89935047 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-935-047 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-935-047 Supplier Abnova Corporation Supplier No. MAB12951
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human TPX2.

Specifications

Antigen TPX2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 18D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human TPX2.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide).
Gene Alias C20orf1/C20orf2/DIL-2/DIL2/FLS353/GD:C20orf1/HCA519/HCTP4/REPP86/p100
Gene Symbols TPX2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-220 of human TPX2.
Purification Method Affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22974
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.