Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582192 Supplier Invitrogen™ Supplier No. PA582192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QELEKSMKMQ QEVVEMRKKN EEFKKLALAG IGQPVKKSVS QVTKSVDFHF RTDERIKQHP KNQEEYKEVN FTSELRKHPS SPARVTKGCT IVKPFNLSQG KKRTFDETVS TYVPLAQQVE DFHKRTPNRY HLRSKKD Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 86%.

The protein encoded by this gene is found in the nucleolus and is similar to that of a family of genes on the Y-chromosome. This gene is intronless. Defects in this gene are a cause of sudden infant death with dysgenesis of the testes syndrome (SIDDT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPX2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tpx2
Gene Accession No. Q9ULW0
Gene Alias 2610005B21Rik; C20orf1; C20orf2; Differentially expressed in cancerous and non-cancerous lung cells 2; differentially expressed in lung cells; DIL2; DIL-2; FLS353; GD: HCA519; GD:C20orf1; HCA519; HCTP4; Hepatocellular carcinoma-associated antigen 519; hepatocellular carcinoma-associated antigen 90; p100; preferentially expressed in colorectal cancer; protein fls353; REPP86; restricted expression proliferation associated protein 100; restricted expression proliferation-associated protein 100; Targeting protein for Xklp2; TPX 2; Tpx2; TPX2 microtubule nucleation factor; TPX2, microtubule nucleation factor; TPX2, microtubule-associated; TPX2, microtubule-associated protein homolog; TPX2, microtubule-associated protein homolog (Xenopus laevis); TPX2, microtubule-associated, homolog
Gene Symbols Tpx2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TPX2. Recombinant protein control fragment (Product #RP-102182).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22974
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.