Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TR alpha/NR1A1/Thyroid Hormone Receptor alpha Antibody (H43), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279822
Description
TR alpha/NR1A1/Thyroid Hormone Receptor alpha Monoclonal specifically detects TR alpha/NR1A1/Thyroid Hormone Receptor alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
TR alpha/NR1A1/Thyroid Hormone Receptor alpha | |
Monoclonal | |
1 mg/mL | |
Western Blot, ELISA | |
AR7, c-ERBA-1, c-erbA-alpha, EAR7, EAR-7, EAR-7.1/EAR-7.2, ERBA, ERBA1thyroid hormone receptor, alpha (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog), ERBA-related 7, MGC000261, MGC43240, NR1A 1, NR1A1THRA3, Nuclear receptor subfamily 1 group A member 1, THRA1ERB-T-1, THRA2, thyroid hormone receptor alpha, thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a)oncogene homolog, avian), triiodothyronine receptor, V-erbA-related protein 7 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at -20 °C. | |
IgG2a |
Western Blot, ELISA | |
H43 | |
Unconjugated | |
PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide | |
THRA | |
Synthetic peptide from the full length Human Thyroid hormone receptor protein | |
0.1 mg | |
Apoptosis, Cancer, GPCR, Neuroscience | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction