Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TR alpha/NR1A1/Thyroid Hormone Receptor alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TR alpha/NR1A1/Thyroid Hormone Receptor alpha |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TR alpha/NR1A1/Thyroid Hormone Receptor alpha Polyclonal specifically detects TR alpha/NR1A1/Thyroid Hormone Receptor alpha in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TR alpha/NR1A1/Thyroid Hormone Receptor alpha | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
AR7, c-ERBA-1, c-erbA-alpha, EAR7, EAR-7, EAR-7.1/EAR-7.2, ERBA, ERBA1thyroid hormone receptor, alpha (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog), ERBA-related 7, MGC000261, MGC43240, NR1A 1, NR1A1THRA3, Nuclear receptor subfamily 1 group A member 1, THRA1ERB-T-1, THRA2, thyroid hormone receptor alpha, thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a)oncogene homolog, avian), triiodothyronine receptor, V-erbA-related protein 7 | |
THRA | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Apoptosis, Cancer, GPCR, Neuroscience | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
7067 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:AFEHYVNHRKHNIPHFWPKLLMKEREVQSSILYKGAAAEGRPGGSLGVHPEGQQLLGMHVVQGPQVRQLEQQLGEAGSLQGPVLQHQSPKSPQQRLLELLHRSGILHARAVCGEDDSSE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title