Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TR alpha/NR1A1/Thyroid Hormone Receptor alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222523
Description
TR alpha/NR1A1/Thyroid Hormone Receptor alpha Polyclonal specifically detects TR alpha/NR1A1/Thyroid Hormone Receptor alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TR alpha/NR1A1/Thyroid Hormone Receptor alpha | |
Polyclonal | |
Unconjugated | |
PBS with 1 mg/ml BSA. with 0.05% Sodium Azide | |
THRA | |
Synthetic Peptide: S(455) R A V C G E D D S S E A S S(469) | |
200 μg | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:100, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
AR7, c-ERBA-1, c-erbA-alpha, EAR7, EAR-7, EAR-7.1/EAR-7.2, ERBA, ERBA1thyroid hormone receptor, alpha (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog), ERBA-related 7, MGC000261, MGC43240, NR1A 1, NR1A1THRA3, Nuclear receptor subfamily 1 group A member 1, THRA1ERB-T-1, THRA2, thyroid hormone receptor alpha, thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a)oncogene homolog, avian), triiodothyronine receptor, V-erbA-related protein 7 | |
Rabbit | |
Affinity Purified | |
RUO | |
7067 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction