Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TR2/NR2C1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171803AF488
Description
TR2/NR2C1 Polyclonal antibody specifically detects TR2/NR2C1 in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
TR2/NR2C1 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
nuclear receptor subfamily 2 group C member 1, nuclear receptor subfamily 2, group C isoform, nuclear receptor subfamily 2, group C, member 1, Orphan nuclear receptor TR2, Testicular receptor 2, TR2, TR2 nuclear hormone receptor, TR2-11 | |
The immunogen for this product maps to a region between residue 1 and 50 of human Testicular Receptor 2 using the numbering given in entry NP_003288.2 (GeneID 7181). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
7181 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction