Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TR2/NR2C1 Antibody (OTI4C7), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274576R
Description
TR2/NR2C1 Monoclonal specifically detects TR2/NR2C1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
TR2/NR2C1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NR2C1 | |
Recombinant protein fragment corresponding to amino acids 1-237 of human TR2/NR2C1 (NP_001027458) produced in E.coli. | |
0.1 mL | |
7181.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI4C7 | |
Western Blot | |
nuclear receptor subfamily 2 group C member 1, nuclear receptor subfamily 2, group C isoform, nuclear receptor subfamily 2, group C, member 1, Orphan nuclear receptor TR2, Testicular receptor 2, TR2, TR2 nuclear hormone receptor, TR2-11 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction