Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527839
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (82%), Rat (81%), Chimpanzee (97%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a nuclear hormone receptor characterized by a highly conserved DNA binding domain (DBD), a variable hinge region, and a carboxy-terminal ligand binding domain (LBD) that is typical for all members of the steroid/thyroid hormone receptor superfamily. This protein also belongs to a large family of ligand-inducible transcription factors that regulate gene expression by binding to specific DNA sequences within promoters of target genes. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these variants has not been determined.
Specifications
TR2 | |
Polyclonal | |
Unconjugated | |
Nr2c1 | |
4831444H07Rik; 80.3; 80-3 cNDA; ATAR; CD270; CD40-like protein; early embryonic nuclear receptor; Eenr; herpes virus entry mediator A; Herpesvirus entry mediator A; HVEA; HVEM; LIGHTR; mTR2; nr2c1; Nuclear receptor subfamily 2 group C member 1; nuclear receptor subfamily 2, group C isoform; nuclear receptor subfamily 2, group C, member 1; nuclear receptor subfamily 2, group H, member 1; orphan nuclear receptor TR2; orphan receptor, TR2-11; pregnancy specific beta-1-glycoprotein 4; Psg4; testicular receptor 2; TNF receptor superfamily member 14; TNFRSF14; Tr2; TR2 nuclear hormone receptor; Tr2-11; Tumor necrosis factor receptor superfamily member 14; tumor necrosis factor receptor superfamily, member 14 (herpesvirus entry mediator); Tumor necrosis factor receptor-like 2; tumor necrosis factor receptor-like gene2; UNQ329/PRO509 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7181 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.8 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P13056 | |
Nr2c1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 419 of Human TR2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction