Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TR4/NR2C2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219579
Description
TR4/NR2C2 Polyclonal specifically detects TR4/NR2C2 in Human samples. It is validated for Western Blot.Specifications
TR4/NR2C2 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
NR2C2 | |
Recombinant protein encompassing a sequence within the center region of human NR2C2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Species reactivity based on sequence identity: Zebrafish (80%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.7mg/mL | |
Western Blot 1:500-1:3000 | |
hTAK1, Nuclear hormone receptor TR4, nuclear receptor subfamily 2 group C member 2, nuclear receptor subfamily 2, group C, member 2, Orphan nuclear receptor TAK1, Orphan nuclear receptor TR4, TAK1TR4TR2R1, Testicular receptor 4, TR4 nuclear hormone receptor | |
Rabbit | |
65 kDa | |
0.1 mL | |
Neuroscience | |
7182 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction