Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TR4/NR2C2 Antibody (SR2096), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32264450UL
Description
TR4/NR2C2 Monoclonal antibody specifically detects TR4/NR2C2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
TR4/NR2C2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR2096 | |
Western Blot 1:500-1:2000 | |
hTAK1, Nuclear hormone receptor TR4, nuclear receptor subfamily 2 group C member 2, nuclear receptor subfamily 2, group C, member 2, Orphan nuclear receptor TAK1, Orphan nuclear receptor TR4, TAK1TR4TR2R1, Testicular receptor 4, TR4 nuclear hormone receptor | |
A synthesized peptide derived from human TR4/NR2C2 (Uniprot #: P49116) | |
50 μg | |
Neuroscience | |
7182 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction