Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRA-1-60 Monoclonal Antibody (TRA-1-60)
Mouse Monoclonal Antibody
$226.00 - $507.00
Specifications
Antigen | TRA-1-60 |
---|---|
Clone | TRA-1-60 |
Concentration | 1 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
Description
TRA-1-60 Monoclonal Antibody for Western Blot, ICC/IF, Flow
TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.Specifications
TRA-1-60 | |
1 mg/mL | |
Flow Cytometry, Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 1 mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
Human embryonal carcinoma cell line 2102Ep. | |
Antibody |
TRA-1-60 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Monoclonal | |
Liquid | |
IgM | |
Human | |
TRA160; tra60 | |
Primary | |
Affinity chromatography - MBP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title