Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRA-1-60(R) Antibody (TRA-1-60), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $487.50
Specifications
Antigen | TRA-1-60(R) |
---|---|
Clone | TRA-1-60 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:200, ELISA reported in scientific literature (PMID 30617132), Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 33234532) |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100730SS
![]() |
Novus Biologicals
NB100730SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB100730
![]() |
Novus Biologicals
NB100730 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
TRA-1-60(R) Monoclonal specifically detects TRA-1-60(R) in Human, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
TRA-1-60(R) | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cellular Markers, Embryonic Stem Cell Markers, Stem Cell Markers | |
RA160, tra61 | |
IgM | |
IgM purified | |
Specific for the neuraminidase resistant form of the TRA-1-60 antigen that is expressed upon the surface of human teratocarcinoma stem cells (EC), human embryonic germ cells (EG) and human embryonic stem cells (ES). TRA-1-60 reacts with a sialidase-sensitive epitope. |
TRA-1-60 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:200, ELISA reported in scientific literature (PMID 30617132), Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin reported in scientific literature (PMID 33234532) | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse (Negative) | |
Human embryonal carcinoma cell line 2102Ep. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title