Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRA-1-81 Antigen Mouse anti-Human, PE, Clone: TRA-1-81, BD

Catalog No. BDB560161 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560161 100 Tests
BDB560885 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560161 Supplier BD Biosciences Supplier No. 560161
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TRA-1-81 monoclonal antibody reacts with a pluripotent-stem-cell-specific epitope on a high-molecular-weight transmembrane glycoprotein. The TRA-1-81 antigen is an epitope on the same keratan sulfate core molecule, podocalyxin, as 4 other distinct antigens on tumor-derived cell lines, TRA-1-60, GCTM2, K4, and K21. The expression of TRA-1-81 antigen is stage-specific and can be used to characterize embryonic cells and monitor their differentiation. The antigen is found on teratocarcinoma (embryonal carcinoma or EC), embryonic inner cell mass (but not morula or trophoblast), and embryonic stem (ES) cells. As human EC and ES cells undergo differentiation, expression of TRA-1-81 antigen is lost.

Host Species: Mouse
Clone: TRA-1-81
Isotype: IgM, κ
Species Reactivity: Human
Immunogen: Human Embryonal Carcinoma Cell Line

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen TRA-1-81 Antigen
Applications Flow Cytometry
Classification Monoclonal
Clone TRA-1-81
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Embryonal Carcinoma Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.