Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRA-1-81 Monoclonal Antibody (TRA-1-81), DyLight™ 550
GREENER_CHOICE

Catalog No. MA1024D550
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1024D550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1024D550 Supplier Invitrogen™ Supplier No. MA1024D550
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-024-D550 has been successfully used in ICC/IF applications on human samples.

TRA-1-81 is a cell surface antigen expressed along with SSEA-3, SSEA-4 and TRA-1-60 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are down-regulated during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRA-1-81
Applications Immunocytochemistry
Classification Monoclonal
Clone TRA-1-81
Concentration 1 mg/mL
Conjugate DyLight 550
Formulation PBS with proprietary stabilizer and 0.02% sodium azide
Gene Alias GCTM-2 antigen; Gp200; PC; PCLP; PCLP1; PCLP-1; Podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; PODXL; TRA180; tra1-81
Host Species Mouse
Immunogen human embryonal carcinoma cell line 2102Ep.
Purification Method Affinity chromatography - MBP
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.