Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRA-1-81 Mouse anti-Human, Alexa Fluor 647, Clone-TRA-1-81, R&D Systems™

Catalog No. IC8495R050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC8495R050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC8495R050 Supplier R&D Systems Supplier No. IC8495R050UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRA-1-81 Monoclonal specifically detects TRA-1-81 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen TRA-1-81
Applications Immunocytochemistry
Classification Monoclonal
Clone TRA-1-81
Conjugate Alexa Fluor 647
Dilution Immunocytochemistry 5-15 ug/mL
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Alias gp200, MGC138240, PC, PCLP, PODXL, TRA181
Host Species Mouse
Immunogen Human embryonal carcinoma cell line 2102Ep
Purification Method IgM-specific Affinity-purified from hybridoma culture supernatant
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human TRA-1-81 in flow cytometry.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.