Learn More
                                    TRA-1-85/CD147 Antibody (TRA-1-85), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3195MFV450
Description
TRA-1-85/CD147 Monoclonal antibody specifically detects TRA-1-85/CD147 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, KnockDownSpecifications
| TRA-1-85/CD147 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| 2120Ep human embryonal carcinoma cell line | |
| RUO | |
| Human | |
| IgG1 | 
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, KnockDown | |
| TRA-1-85 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | 