Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRA16 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980217
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0217 Supplier Bioss Supplier No. BS0329R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May act as a repressor of NR2C2-mediated transactivation by suppressing the binding between NR2C2/TR4 and the TR4-response element in target genes.

Specifications

Antigen TRA16
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NR2C2AP
Gene Accession No. Q86WQ0
Gene Alias NR2C2AP; nuclear receptor 2C2 associated protein; Nuclear receptor 2C2-associated protein; repressor for TR4 transactivation; TR4 orphan receptor associated protein TRA16; TR4 orphan receptor-associated 16 kDa protein; TRA16
Gene Symbols NR2C2AP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human of human TRA16.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126382
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.