Learn More
TRA2A Polyclonal Antibody

Description
TRA2A Polyclonal Antibody for ICC/IF, IHC
Specifications
Specifications
| Antigen | TRA2A |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TRA2A |
| Gene Accession No. | Q13595 |
| Gene Alias | 1500010G04Rik; AL022798; AWMS1; G430041M01Rik; HSU53209; htra-2 alpha; htra-2-alpha; mAWMS1; nuclear transformer-2 alpha; putative MAPK activating protein PM24; RGD1562563; TRA-2 alpha; TRA2A; Tra2alpha; TRA2-alpha; transformer 2 alpha homolog; transformer 2 alpha homolog (Drosophila); transformer-2 alpha; transformer-2 protein homolog A; Transformer-2 protein homolog alpha |
| Gene Symbols | TRA2A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.