Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRA2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575290
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
TRA2A (Transformer 2 alpha) is a member of the transformer 2 homolog family and encodes a protein with two RS domains and an RRM (RNA recognition motif) domain. It is a sequence specific RNA binding protein which participates in the regulation of pre-mRNA splicing.
Specifications
TRA2A | |
Polyclonal | |
Unconjugated | |
TRA2A | |
1500010G04Rik; AL022798; AWMS1; G430041M01Rik; HSU53209; htra-2 alpha; htra-2-alpha; mAWMS1; nuclear transformer-2 alpha; putative MAPK activating protein PM24; RGD1562563; TRA-2 alpha; TRA2A; Tra2alpha; TRA2-alpha; transformer 2 alpha homolog; transformer 2 alpha homolog (Drosophila); transformer-2 alpha; transformer-2 protein homolog A; Transformer-2 protein homolog alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
101214, 29896, 500116 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13595, Q6PFR5 | |
TRA2A | |
Synthetic peptide corresponding to amino acids 210-258 of Human TRA2A. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction