Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRA2B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157456
Description
TRA2B Polyclonal specifically detects TRA2B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TRA2B | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
DKFZp686F18120, hTRA2-beta, SFRS10, Splicing factor, arginine/serine-rich 10, splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila), SRFS10, TRA-2 beta, TRA2-beta, TRAN2B, transformer 2 beta homolog (Drosophila), transformer 2 homolog, Transformer-2 protein homolog B, transformer-2 protein homolog beta, transformer-2-beta | |
Rabbit | |
Protein A purified | |
RUO | |
6434 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
P62995 | |
TRA2B | |
Synthetic peptides corresponding to SFRS10 (splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila)) The peptide sequence was selected from the N terminal of SFRS10. Peptide sequence MSDSGEQNYGERESRSASRSGSAHGSGKSARHTPARSRSKEDSRRSRSKS The peptide sequence for this immunogen was taken from within the described region. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Rat: 100%; Chicken: 100%; Sumatran orangutan: 100%; Bovine: 100%; Pig: 100%; Mouse: 100%; Canine: 100%; Human: 100%; Xenopus: 85%; Western clawed frog: 85%;. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction