Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRA2B Polyclonal Antibody, Proteintech
SDP

Catalog No. 502333410 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-3410 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-3410 Supplier Proteintech Group Inc Supplier No. 296271AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRA2B Polyclonal antibody specifically detects TRA2B in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen TRA2B
Applications Western Blot
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Accession No. P62995
Gene Alias 5730405G21Rik; D16Ertd266e; Htra2-beta; PPP1R156; SFRS10; SIG-41; Silg41; SRFS10; TRA2-BETA; TRA2B; TRA2beta; TRAN2B
Host Species Rabbit
Purification Method Antigen affinity chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6434
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.