Learn More
TRA2B/SFRS10 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | TRA2B/SFRS10 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-9278
![]() |
Bethyl Laboratories, Inc
A305011AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
SFRS10 contains 1 RRM (RNA recognition motif) domain and belongs to the splicing factor SR family. It is a sequence-specific RNA-binding protein which participates in the control of pre-mRNA splicing.Specifications
TRA2B/SFRS10 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Htra2-beta, SFRS10, SRFS10, TRA2-BETA, TRAN2B, TRA-2 beta, splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila), transformer-2 protein homolog B, transformer-2 protein homolog beta | |
Between 1 and 50 | |
Primary | |
4° C | |
TRA2B |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P62995, P62996 | |
20462, 6434 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.