Learn More
TRAAK Mouse anti-Mouse,Rat, Unconjugated, Clone: N445/27 , MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | TRAAK |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | N445/27 |
| Conjugate | Unconjugated |
| Gene Accession No. | NP_032457 |
| Gene Alias | Potassium channel subfamily K member 4;TWIK-related arachidonic acid-stimulated potassium channel protein;TRAAK |
| Gene Symbols | Kcnk4, Traak |
| Host Species | Mouse |
| Immunogen | A recombinant fragment corresponding to 143 amino acids from the C-terminal, cytoplasmic domain of mouse Potassium channel subfamily K member 4 (TRAAK). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.