Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAAK Mouse anti-Mouse,Rat, Unconjugated, Clone: N445/27 , MilliporeSigma™

Catalog No. MABN23932MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN23932MI 25 μg
MABN23931MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN23932MI Supplier MilliporeSigma Supplier No. MABN239325UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAAK Monoclonal antibody specifically detects TRAAK in Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAAK
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone N445/27
Conjugate Unconjugated
Gene Accession No. NP_032457
Gene Alias Potassium channel subfamily K member 4;TWIK-related arachidonic acid-stimulated potassium channel protein;TRAAK
Gene Symbols Kcnk4, Traak
Host Species Mouse
Immunogen A recombinant fragment corresponding to 143 amino acids from the C-terminal, cytoplasmic domain of mouse Potassium channel subfamily K member 4 (TRAAK).
Molecular Weight of Antigen 43.05 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone N445/27 is a mouse monoclonal antibody that detects Potassium channel subfamily K member 4 (TRAAK). It targets an epitope within the C-terminal, cytoplasmic domain.
Target Species Mouse, Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.