Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRABD Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542216
Description
Peptide sequence: MDGEEQQPPH EANVEPVVPS EASEPVPRVL SGDPQNLSDV DAFNLLLEMK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.
The function of TRABD remains unknown.
Specifications
| TRABD | |
| Polyclonal | |
| Unconjugated | |
| TRABD | |
| 5730502D15Rik; AL023039; fb66c10; hypothetical protein LOC322581; hypothetical protein PP2447; LP6054; PP2447; Protein TTG2; RGD1310875; TraB domain containing; traB domain-containing protein; TRABD; TTG2; wu:fb66c10; wu:fi04e11; zgc:66436 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 80305 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H4I3 | |
| TRABD | |
| Synthetic peptide directed towards the N-terminal of human PP2447 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction