Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRABD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542217
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542217 Supplier Invitrogen™ Supplier No. PA542217
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LCFLSDPISK DDVERCKQKD LLEQMMAEMI GEFPDLHRTI VSERDVYLTY Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 86%.

The function of TRABD remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRABD
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRABD
Gene Accession No. Q9H4I3
Gene Alias 5730502D15Rik; AL023039; fb66c10; hypothetical protein LOC322581; hypothetical protein PP2447; LP6054; PP2447; Protein TTG2; RGD1310875; TraB domain containing; traB domain-containing protein; TRABD; TTG2; wu:fb66c10; wu:fi04e11; zgc:66436
Gene Symbols TRABD
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TRABD.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80305
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.