Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAC-1 Monoclonal Antibody (1D3)
GREENER_CHOICE

Catalog No. PIMA522082
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522082 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522082 Supplier Invitrogen™ Supplier No. MA522082
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: FCQRELYEDS LLDHCITHHR SERRPVFCPL CRLIPDENPS SFSGSLIRHL QVSHTLFYDD FIDFNIIEEA LIRRVLDRSL LEYVNHSNT.

TRAC-1 is a 232 amino acid protein containing an N-terminal RING-type zinc finger domain. It functions as a positive regulator in the T-cell receptor signaling pathway, thus leading to T-cell activation. It mediates ubiquitination and subsequent proteasomal degradation of target proteins. TRAC-1 is expressed in lymphoid tissues, including bone marrow, spleen, thymus as well as in other tissues. It is predominantly expressed in the CD4+ and CD8+ T-cells, suggesting that it is preferentially confined to T-cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAC-1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1D3
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RNF125
Gene Accession No. Q96EQ8
Gene Alias 4930553F04Rik; E3 ubiquitin-protein ligase RNF125; RING finger protein 125; ring finger protein 125, E3 ubiquitin protein ligase; RNF125; T-cell RING activation protein 1; T-cell ring protein identified in activation screen; TNORS; TRAC1; TRAC-1
Gene Symbols RNF125
Host Species Mouse
Immunogen RNF125 (NP_060301, 143 a.a. ~ 231 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54941
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.