Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAC-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559607
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559607 Supplier Invitrogen™ Supplier No. PA559607
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CIATSLKNNK WTCPYCRAYL PSEGVPATDV AKRMKSEYKN CAECDTLVCL SEMRAHIRTC QKYIDKYGPL QELEETAARC VCP Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 88%.

TRAC-1 is a 232 amino acid protein containing an N-terminal RING-type zinc finger domain. It functions as a positive regulator in the T-cell receptor signaling pathway, thus leading to T-cell activation. It mediates ubiquitination and subsequent proteasomal degradation of target proteins. TRAC-1 is expressed in lymphoid tissues, including bone marrow, spleen, thymus as well as in other tissues. It is predominantly expressed in the CD4+ and CD8+ T-cells, suggesting that it is preferentially confined to T-cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAC-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF125
Gene Accession No. Q96EQ8
Gene Alias 4930553F04Rik; E3 ubiquitin-protein ligase RNF125; RING finger protein 125; ring finger protein 125, E3 ubiquitin protein ligase; RNF125; T-cell RING activation protein 1; T-cell ring protein identified in activation screen; TNORS; TRAC1; TRAC-1
Gene Symbols RNF125
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRAC-1. Recombinant protein control fragment (Product #RP-98228).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54941
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.