Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRAC-1 Polyclonal Antibody
TRAC-1 Polyclonal Antibody
Supplier: Thermo Scientific PA590268
Description
TRAC-1 Polyclonal Antibody for Western Blot

Specifications
TRAC-1 | |
Polyclonal | |
Unconjugated | |
RNF125 | |
4930553F04Rik; E3 ubiquitin-protein ligase RNF125; RING finger protein 125; ring finger protein 125, E3 ubiquitin protein ligase; RNF125; T-cell RING activation protein 1; T-cell ring protein identified in activation screen; TNORS; TRAC1; TRAC-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54941 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.97 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q96EQ8 | |
RNF125 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human RNF125 (NP_0603012). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction