Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trace Amine Receptor 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Specifications
Antigen | Trace Amine Receptor 1 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224714SS
![]() |
Novus Biologicals
NBP224714SS |
0.025 mg | N/A | N/A | N/A | ||||
NBP224714
![]() |
Novus Biologicals
NBP224714 |
0.1 mg | N/A | N/A | N/A | ||||
Description
Trace Amine Receptor 1 Polyclonal specifically detects Trace Amine Receptor 1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Trace Amine Receptor 1 | |
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Primate | |
Q96RJ0 | |
134864 | |
A portion of amino acids 200-230 of human TAAR1 was used as the immunogen | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS with 0.05% Sodium Azide | |
MGC126874, MGC138399, TA1RP11-295F4.9, TaR-1, TAR1trace amine-associated receptor 1, trace amine associated receptor 1, Trace amine receptor 1taR-1, TRAR1 | |
TAAR1 | |
IgG | |
Protein G purified | |
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 93% homologous in rhesus monkey, 86% homologous in dog and 50% homologous in mouse and rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title