Learn More
Fisherbrand™ Traceable™ 7600 Smart Probes
Remove the stress of recalibration
 View more versions of this product
Catalog No. 01-257-639
Only null left
Add to Cart
Description
Description
  • Plug-and-Play Smart Probes store calibration and measurement limits for quick set-up
  • The 7600-series data logger will auto recognize the probes' parameters
  • When it's time to recalibrate, easily swap the probe for a new one
  • Allows custom configurations for every data logger
  • A door gasket can be closed on the probe cables for use in refrigerators and freezers
7600-Series Smart Probes are calibrated in place of the base data logger. They remove downtime in the lab by allowing an outdated sensor to be quickly swapped out with a new pre-calibrated one – no need to worry about sending the data logger for recalibration. Outdated sensors can then be disposed of or recalibrated without losing any valuable time.
Traceable™ Products have an individually numbered Traceable™ Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Certificates are guaranteed to have at least one year of calibration remaining.
Specifications
Specifications
Probe
1 Year
-50°C to 60°C
Bullet
-58°F to 140°F
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.