Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ 1-Methylnaphthalene Solution, 2,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020271
Click to view available options
Quantity:
1 mL

1-Methylnaphthalene is an oily liquid having an earthy phenolic type of odor. It is an important constituent of polycyclic aromatic hydrocarbons, which can be commonly obtained from coal and petroleum oils.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line TraceCERT
CAS 90-12-0
Synonym alpha-Methylnaphthalene
Type Organic Pollutant Standard
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 2,000 μg/mL in methanol

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.