Learn More
TraceCERT™ Heavy Metal Mix, Certified Reference Material, MilliporeSigma™ Supelco™
Catalog No. 111022435 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
100 mL
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.
  • According to USP <561> Articles of Botanical Origin
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL
Grade TraceCERT™ Analytical Standard
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.