Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TraceCERT™ Indoor Air Standard 50 components, 100 μg/mL each component in Methanol: Water (19:1), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
1 mL
3 x 1 mL
Specifications
Specifications
Packaging | Ampule |
Product Line | TraceCERT |
CAS | 67-56-1 |
Flash Point | 50°C |
Type | Certified Reference Material |
Quantity | 1 mL |
Grade | TraceCERT™ Analytical Standard |
Concentration or Composition (by Analyte or Components) | 100 μg/mL each component in methanol: water (19:1) |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction