Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TraceGOLD TG-POLAR GC Columns

Catalog No. 260825080
Change view
Click to view available options
Length (Metric):
30 m
60 m
105 m
Diameter (Metric) Inner:
0.25 mm
0.53 mm
Film Thickness:
0.1 μm
0.2 μm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Inner Film Thickness
26-082-5080 60 m 0.53 mm 0.2 μm
26-082-5000 105 m 0.25 mm 0.1 μm
26-082-5010 30 m 0.25 mm 0.2 μm
26-082-5020 60 m 0.25 mm 0.2 μm
26-082-5030 105 m 0.25 mm 0.2 μm
26-082-5070 30 m 0.53 mm 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 26-082-5080 Supplier Thermo Scientific™ Supplier No. 260825080

Redefine your expectations for high-polarity GC columns with the Thermo Scientific TraceGold TG-POLAR Columns

Refine your expectations for high-polarity GC/MS with the Thermo Scientific™ TraceGOLD™ TG-POLAR columns, which are specifically designed with a polymer and surface treatment to overcome traditional problems with high-polarity columns.

Engineered for Superior Performance

  • Highly polar phase, 90% biscyanopropyl/10% phenylcyanopropyl polysiloxane, not bonded
  • Strong dipole moment and high selectivity for cis/trans compounds or compounds with conjugated double bonds
  • Equivalent to U.S.P G8 and G48

Applications:

  • Cis/Trans fatty acid methyl esters (FAMEs)
  • Dioxins

A Leap Forward in Performance with Low Bleed and Superior Inertness:
  • A low bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low-level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior intertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Alternative Uses:
  • Alcohols
  • Aldehydes
  • Biodiesel–FAMEs
Similar To:
  • DB-23
  • HP-23
  • Rtx™™-2330
  • SP™-2330
  • SP™-2380
  • SPB™-2560
  • HP-88
  • Silar™™ 90c
  • CP-Sil 88 FAME
  • CP-Sil 88
  • BPX 70
  • BPX 90
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) GC/MS of Cis/Trans FAMEs, GC/MS of Dioxins
Film Thickness 0.2 μm
Length (Metric) 60 m
Polarity Highly Polar
Description 0.2 um Thickness; 0.53 mm ID; 60 m Length
Diameter (Metric) 0.53 mm
USP Type G8, G48
Stationary Phase TraceGOLD™ TG-Polar
Diameter (Metric) Inner 0.53 mm
Max. Temperature (Metric) 260/275°C

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.