Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TraceGOLD™ TG-WaxMS A GC Columns

Catalog No. 260872240
Change view
Click to view available options
Length (Metric):
15 m
30 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.5 μm
1 μm
1.5 μm
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Inner Film Thickness
26-087-2240 30 m 0.32 mm 0.5 μm
26-087-1420 30 m 0.25 mm 0.25 μm
26-087-1430 30 m 0.32 mm 0.25 μm
26-087-1540 60 m 0.25 mm 0.25 μm
26-087-2230 30 m 0.25 mm 0.5 μm
26-087-2970 30 m 0.32 mm 1 μm
26-087-2980 30 m 0.53 mm 1 μm
26-087-2110 15 m 0.25 mm 0.5 μm
26-087-2120 15 m 0.32 mm 0.5 μm
26-087-2130 15 m 0.53 mm 0.5 μm
26-087-2250 30 m 0.53 mm 0.5 μm
26-087-2350 60 m 0.25 mm 0.5 μm
26-087-2360 60 m 0.32 mm 0.5 μm
26-087-2370 60 m 0.53 mm 0.5 μm
26-087-2850 15 m 0.32 mm 1 μm
26-087-2860 15 m 0.53 mm 1 μm
26-087-3090 60 m 0.32 mm 1 μm
26-087-3100 60 m 0.53 mm 1 μm
26-087-3340 15 m 0.53 mm 1.5 μm
26-087-3360 30 m 0.53 mm 1.5 μm
26-087-3370 60 m 0.53 mm 1.5 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 26-087-2240 Supplier Thermo Scientific™ Supplier No. 260872240

Identify organic acids with high performance, sensitivity and reproducibility while preventing oxidative damage with this low-bleed, polar GC column.

Identify organic acids with high performance, sensitivity and reproducibility while preventing oxidative damage using Thermo ScientificTM TraceGOLDTM TG-WaxMS A GC column.

Engineered for Superior Performance
  • Polar phase, acid-deactivated polyethylene glycol
  • Resists oxidative damage and adsorption of acids
  • Excellent peak shapes for high MW acids
  • Equivalent to USP G25, G35 phases
Applications:
  • Organic acids
  • Free fatty acids
  • Alcohols

A Leap Forward in Column Performance with Low Bleed and Superior Inertness
  • This low-bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.

Recommended For:
  • Acids
  • Carboxylic acids
  • Organic acids
  • Alternative Uses:
  • Alcohols
  • Alcohols in beverages
  • Aldehydes
  • Alditol acetates (sugars)
  • Essential oils
  • Similar To:
  • DB-FFAP
  • HP-FFAP
  • Nukol™
  • OV™-351
  • CP-Wax 58 CB
  • Stabilwax™-DA
  • BP-21
  • Optima™ FFAP
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Free Fatty Acids Analyis, GC/MS for Free Fatty Acids, GC/MS for Alcohols
Film Thickness 0.5 μm
Length (Metric) 30 m
Polarity Polar
Description 0.5 um Thickness; 0.32 mm ID; 30 m Length
Diameter (Metric) 0.32 mm
USP Type G25, G35
Stationary Phase TraceGOLD™ TG-WAXMS A
Diameter (Metric) Inner 0.32 mm
Product Line TraceGOLD
Max. Temperature (Metric) 240/250°C
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.