Learn More
Thermo Scientific™ TraceGOLD™ TG-WaxMS A GC Columns
Description
Identify organic acids with high performance, sensitivity and reproducibility while preventing oxidative damage using Thermo ScientificTM TraceGOLDTM TG-WaxMS A GC column.
- Polar phase, acid-deactivated polyethylene glycol
- Resists oxidative damage and adsorption of acids
- Excellent peak shapes for high MW acids
- Equivalent to USP G25, G35 phases
- Organic acids
- Free fatty acids
- Alcohols
A Leap Forward in Column Performance with Low Bleed and Superior Inertness
- This low-bleed, stable column produces consistent results.
- Low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
- Minimizing column bleed extends column lifetime, leading to higher productivity.
- Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
- Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Recommended For:
- Acids
- Carboxylic acids
- Organic acids
- Alternative Uses:
- Alcohols
- Alcohols in beverages
- Aldehydes
- Alditol acetates (sugars)
- Essential oils
- Similar To:
- DB-FFAP
- HP-FFAP
- Nukol™
- OV™-351
- CP-Wax 58 CB
- Stabilwax™-DA
- BP-21
- Optima™ FFAP
Specifications
Specifications
| For Use With (Application) | GC/MS for Organic Acids, GC/MS for Free Fatty Acids, GC/MS for Alcohols |
| Film Thickness | 1 μm |
| Length (Metric) | 60 m |
| Polarity | Polar |
| Description | 1umthickness; 0.32mm ID; 60m length |
| Diameter (Metric) | 0.32 mm |
| USP Type | G25, G35 |
| Stationary Phase | TraceGOLD™ TG-WAXMS A |
| Diameter (Metric) Inner | 0.32 mm |
| Max. Temperature (Metric) | 240/250°C |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.