Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRADD Antibody (313203), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2658F
Description
TRADD Monoclonal antibody specifically detects TRADD in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
TRADD | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
313203 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
MGC11078, TNFRSF1A-associated via death domainTNFR1-associated DEATH domain protein, tumor necrosis factor receptor type 1 associated death domain protein, tumor necrosis factor receptor type 1-associated DEATH domain protein, tumor necrosis factor receptor-1-associated protein | |
E. coli-derived recombinant human TRADD, Met1-Ala312, Accession # Q15628 | |
0.1 mL | |
Apoptosis, Cancer, Death Receptor Signaling Pathway, Tumor Suppressors | |
8717 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction