Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRADD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588134
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588134 Supplier Invitrogen™ Supplier No. PA588134
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, Jurkat; Cellular Location: Cytoplasm, Nucleus, cytoskeleton Immunogen sequence: AQTFLFQGQP VVNRPLSLKD QQTFARSVGL KWRKVGRSLQ RGCRALRDPA LDSLAYEYER EGLYEQAFQL LRRFVQAEGR RATLQRLVEA LEENELTSLA EDLLGLTDPN GGLA.

TRADD (Tumor necrosis factor receptor type 1-associated DEATH domain protein) is an important mediator of FAS and TNFR1 associated apoptotic signaling. The nuclear form acts as a tumor suppressor by preventing ubiquitination and degradation of isoform p19ARF/ARF of CDKN2A by TRIP12. TRADD acts by interacting with TRIP12 and leading to the disruption of the interaction between TRIP12 and isoform p19ARF/ARF of CDKN2A. TRADD acts as an adapter molecule for TNFRSF1A/TNFR1 that specifically associates with the cytoplasmic domain of activated TNFRSF1A/TNFR1 mediating its interation with FADD. Overexpression of TRADD leads to two major TNF-induced responses - apoptosis and activation of NF-kappa-B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRADD
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene TRADD
Gene Accession No. Q15628
Gene Alias 9130005N23Rik; AA930854; Hs.89862; Hs89862; MGC11078; TNF receptor 1 associated signal transducer; TNFR1-associated DEATH domain protein; TNFRSF1A associated via death domain; TNFRSF1A-associated via death domain; TRADD; tumor necrosis factor receptor type 1 associated death domain protein; tumor necrosis factor receptor type 1-associated DEATH domain protein; tumor necrosis factor receptor-1-associated protein
Gene Symbols TRADD
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8717
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.