Learn More
Eppendorf™ Trade-In Research™ Plus
Catalog No. 05414256 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
500 μL
1000 μL
Color:
Blue
Light Gray
Medium Gray
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
05-414-256 100 μL Yellow
05-414-250 10 μL Medium Gray
05-414-251 10 μL Yellow
05-414-252 20 μL Light Gray
05-414-253 20 μL Yellow
05-414-254 25 μL Yellow
05-414-255 50 μL Yellow
05-414-257 200 μL Yellow
05-414-258 200 μL Blue
05-414-259 250 μL Blue
05-414-260 500 μL Blue
05-414-261 1000 μL Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 05-414-256 Supplier Eppendorf™ Supplier No. 2231300027
Add to Cart
Add to Cart
This item is not returnable. View return policy
Trade-In Research Plus pipettes do not have to be in working condition and may be any manufacturers brand.
  • Returned pipettes do not have to be in working condition and may be any manufacturers brand.
  • Pipettes must be free of contamination and returned together with the signed Decontamination Form (www.eppendorf.com/decontamination) to the following address:
    • In U.S.: Eppendorf Services, 151 Memorial Drive, Suite B, Shrewsbury, MA 01545
    • In Canada: Eppendorf Canada, 2810 Argentia Road, Unit #2, Mississauga, ONT L5N 8L2
  • If you have any questions, please contact Customer Support at:
    • U.S.: 800-645-3050
    • Canada: 800-263-8715
  • Benefit from one of the lowest operating and tip ejection forces on the pipette market, helping to reduce hand and arm strain
  • Temporarily adjust your pipette in seconds for better accuracy when pipetting challenging liquids like ethanol or when pipetting at high altitudes. Return to factory adjustment without calibration
  • Autoclave the entire pipette or only the lower part according to your needs and applications to ensure a proper decontamination
  • Premium material ensures a high UV and chemical resistance
  • Enjoy excellent flexibility and choose among single-channel pipettes in fixed or variable volume as well as 8-, 12-, 16- and 24-channel pipettes
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Yellow
Number of Channels 1
Type Pipette
Volume (Metric) 100 μL